missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ ID Tape

Product Code. 12774758 Shop All Sampling Systems Products
Click to view available options
Display Text:
Quarantine
Rejected
Released
Sampled
Unit Size:
10 rolls
Pack of 10
This item is not returnable. View return policy

Product Code. 12774758

Brand: Sampling Systems™ 8221A02

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Quickly identify the status of a container to prevent it being used in error. Sampling Systems™ ID Tape allows containers, pallets, and bags to be easily labelled with highly visible tape.

  • Use with standard tape dispensers
  • High strength adhesive
  • 10 rolls per box

Specifications

Description Medium strength adhesion tape
Color Red
Product Type Labeled ID Tape
Display Text Sampled
No. per Pack 10/Pk.
Dimensions (LxW) 66 x 50 mm
Length (Metric) 66 mm
Width (Metric) 50 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.