missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IDE 1
GREENER_CHOICE

Product Code. 18084142
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084142 50 mg 50mg
18016032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084142 Supplier Tocris Bioscience™ Supplier No. 4015/50
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Induces definitive endoderm formation in mouse and human ESCs

Induces definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 125 nM). Thought to activate the TGF-β signaling pathway; induces Smad2 phosphorylation and increases levels of Nodal expression.

This compound is mixture of E/Z isomers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1160927-48-9
Molecular Formula C15H18N2O5

Specifications

Quantity 50 mg
Formula Weight 306.31
Purity >98%
Molecular Weight (g/mol) 306.318
Chemical Name or Material IDE 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.