missing translation for 'onlineSavingsMsg'
Learn More

Idebenone, MedChemExpress

Product Code. 16499046
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499046 100 mg 100mg
16418948 10 mg 10mg
16404817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499046 Supplier Medchem Express Supplier No. HYN0303
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Idebenone, a well-appreciated mitochondrial protectant, exhibits protective efficacy against neurotoxicity and can be used for the research of Alzheimer's disease, Huntington's disease.

Idebenone (oxidised form) has a dose-dependent inhibitory effect on the enzymatic metabolism of arachidonic acid in astroglial homogenates (IC50=16.65 μM). Idebenone, a coenzyme Q10 analog and an antioxidant, induces apoptotic cell death in the human dopaminergic neuroblastoma SHSY-5Y cells. Idebenone quickly crosses the blood-brain barrier.

Specifications

Chemical Name or Material Idebenone
CAS 58186-27-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H30O5
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (295.47 mM)
SMILES O=C1C(CCCCCCCCCCO)=C(C)C(C(OC)=C1OC)=O
Molecular Weight (g/mol) 338.44
Formula Weight 338.44
Percent Purity 97.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.