missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Idelalisib
GREENER_CHOICE

Product Code. 18777016
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18777016 50 mg 50mg
18749626 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18777016 Supplier Tocris Bioscience™ Supplier No. 7631/50
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent PI 3-Kinase δ isoform inhibitor

Idelalisib is a potent PI 3-Kinase δ inhibitor (IC50 = 2.5 nM for p110δ); it exhibits >40-fold selectivity for the δ isoform over other PI 3-K class I enzymes (IC50s = 89, 565, and 820 nM for p110γ, p110β, and p110α respectively). Idelalisib blocks constitutive PI 3-K signaling in malignant B-cell lines and primary patient tumor cells. Idelalisib inhibits chronic lymphocytic leukemia cell chemotaxis and downregulates chemokine secretion triggered by B-cell receptor signaling in stromal cocultures. Idelalisib lacks significant cytotoxicity toward natural killer (NK) cells. Idelalisib also decreases production of inflammatory cytokines, including TNF-γ, CD40L, and IL-6 by T cells and IFN-γ by NK cells. Idelalisib is cell-permeable and orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluoro-3-phenyl-2-[(1 S)-1-(9 H-purin-6-ylamino)propyl]-4(3 H)-quinazolinone
CAS 870281-82-6
Quantity 50 mg
Target PI 3-Kinase Inhibitors
Molecular Formula C22H18FN7O
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.