missing translation for 'onlineSavingsMsg'
Learn More

Idoxuridine, MedChemExpress

Product Code. 16470868
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470868 5 g 5g
16462129 1 g 1g
16418448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16470868 Supplier Medchem Express Supplier No. HYB0307
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.

Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Idoxuridine
CAS 54-42-2
Color White
Purity Grade Notes Research
Molecular Formula C9H11IN2O5
Quantity 5 g
Synonym 5-Iodo-2′-deoxyuridine 5-IUdR IdUrd
Solubility Information DMSO : 125 mg/mL (353.01 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(I)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 354.1
Formula Weight 354.1
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.