missing translation for 'onlineSavingsMsg'
Learn More

Idramantone, MedChemExpress

Product Code. 16468576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468576 Supplier Medchem Express Supplier No. HYB1044
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Idramantone (Kemantane), an Adamantane derivative, is an immunostimulant.

Idramantone (Kemantane), an Adamantane derivative, is an immunostimulant.

Specifications

Chemical Name or Material Idramantone
CAS 20098-14-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H14O2
Quantity 100 mg
Synonym Kemantane 5-Hydroxy-2-adamantanone
Solubility Information H2O : 20 mg/mL (120.32 mM; Need ultrasonic)
SMILES O=C1C(C2)CC3CC2(O)CC1C3
Molecular Weight (g/mol) 166.22
Formula Weight 166.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.