missing translation for 'onlineSavingsMsg'
Learn More

Ifenprodil hemitartrate, 99%

Product Code. 15443619
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443619 10 mg 10mg
15453619 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443619 Supplier Thermo Scientific Alfa Aesar Supplier No. J60932.MA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

NMDA receptor antagonist, acting at the polyamine site | CAS: 23210-56-2 | C21H27NO2 | 325.452 g/mol

NMDA receptor antagonist, acting at the polyamine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23210-56-2
Molecular Formula C21H27NO2
Molecular Weight (g/mol) 325.452
MDL Number MFCD01529999
InChI Key UYNVMODNBIQBMV-UHFFFAOYSA-N
Synonym ifenprodil, vadilex, ifenprodil tartrate, dilvax, creocral, ifenprodil inn:dcf, ifenprodilum inn-latin, unii-r8oe3p6o5s, 4-2-4-benzylpiperidin-1-yl-1-hydroxypropyl phenol, r8oe3p6o5s
PubChem CID 3689
IUPAC Name 4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol
SMILES CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3

Specifications

Color White
Quantity 10 mg
Merck Index 14,4895
Formula Weight 400.49
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ifenprodil hemitartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.