missing translation for 'onlineSavingsMsg'
Learn More

Ifenprodil, TRC

Product Code. 30394172
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394172 50 mg 50mg
30390009 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394172 Supplier Toronto Research Chemicals Supplier No. TRCI25800350MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23210-56-2
Molecular Formula C21H27NO2
Molecular Weight (g/mol) 325.44
Synonym alpha-(4-Hydroxyphenyl)-beta-methyl-4-(phenylmethyl)-1-piperidineethanol, (±)-Ifenprodil, 1-Methyl-2-hydroxy-2-(4-hydroxyphenyl)ethyl-1-(4-benzylpiperidine), 2-(4-Benzylpiperidino)-1-(1-hydroxyphenyl)-1-propanol, RC 61-91
IUPAC Name 4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol
SMILES CC(C(O)c1ccc(O)cc1)N2CCC(Cc3ccccc3)CC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H27NO2/c1-16(21(24)19-7-9-20(23)10-8-19)22-13-11-18(12-14-22)15-17-5-3-2-4-6-17/h2-10,16,18,21,23-24H,11-15H2,1H3
Formula Weight 325.2042
Chemical Name or Material Ifenprodil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.