Learn More
Invitrogen™ Rat IFN gamma ELISA Kit
Description
The Rat Interferon gamma (Rt IFNγ) ELISA quantitates Rt IFNγ in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt IFNγ. Principle of the method The Rat IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P01581 |
| Assay Range | 8-500 pg/mL |
| Assay Sensitivity | <2 pg/mL |
| Content And Storage | Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Description | IFN gamma ELISA Kit, 1 Kit, 480 Assays, 50μL Sample Size, 8 to 500pg/mL Assay Range, 2.5 hrs. Assay Time, Rat Species, IFNγ Target, HRP Conjugate, TMB Substrate, < 2pg/mL Sensitivity, Colorimetric Detection Method, Store kit at 2 to 8°C, Used fo |
| Assay | 480 |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.