missing translation for 'onlineSavingsMsg'
Learn More

Ifosfamide

Product Code. 15628735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15628735 1 g 1g
15638735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15628735 Supplier Thermo Scientific Acros Supplier No. 461350010
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3778-73-2 | C7H15Cl2N2O2P | 261.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3778-73-2
Molecular Formula C7H15Cl2N2O2P
Molecular Weight (g/mol) 261.09
InChI Key HOMGKSMUEGBAAB-UHFFFAOYSA-N
Synonym ifosfamide, isophosphamide, iphosphamide, isofosfamide, ifosfamid, mitoxana, naxamide, iphosphamid, isoendoxan, ifex
PubChem CID 3690
ChEBI CHEBI:5864
IUPAC Name N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine
SMILES C1CN(P(=O)(OC1)NCCCl)CCCl

Specifications

Melting Point 39.0°C to 42.0°C
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 261.09
Percent Purity ≥97.5% (ELSD) (HPLC)
Chemical Name or Material Ifosfamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.