missing translation for 'onlineSavingsMsg'
Learn More

Ifosfamide, TRC

Product Code. 30136598
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136598 100 mg 100mg
18013940 250 mg 250mg
18063330 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136598 Supplier Toronto Research Chemicals Supplier No. TRCI265000100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3778-73-2
Molecular Formula C7 H15 Cl2 N2 O2 P
Molecular Weight (g/mol) 261.09
Synonym Ifosfamide, (RS)-N,3-bis(2-chloroethyl)-1,3,2-oxazaphosphinan-2-amine 2-oxide
IUPAC Name N,3-bis(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine
SMILES ClCCNP1(=O)OCCCN1CCCl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H15Cl2N2O2P/c8-2-4-10-14(12)11(6-3-9)5-1-7-13-14/h1-7H2,(H,10,12)
Formula Weight 260.0248
Chemical Name or Material Ifosfamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.