Learn More
IgA (α) Goat anti-Mouse, HRP, Secondary Antibody, Proteintech
Goat anti-Mouse Affinipure Secondary Antibody
Brand:  Proteintech SA00012-7-200UL
 View more versions of this product
Product Code. 17610397
  • 50.00 € / 200µL
Specifications
IgA (α)
HRP
Goat
200 μL
Mouse
Liquid
Western Blot
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.