missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD30/TNFRSF8 Fc Avi-tag Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
When Recombinant Human CD30 Ligand/TNFSF8 is immobilized at 0.25 μg/mL (100 μL/well), the concentration of Biotinylated Recombinant Human CD30/TNFRSF8 Fc Chimera Avi-tag that produces 50% of the optimal binding response is 3 to 15 ng/mL.
Specifications
Specifications
| Accession Number | P29808.1 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 943 |
| Molecular Weight (g/mol) | 107 to 127 kDa, under reducing conditions |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD30/TNFRSF8 protein, (N-terminus) Human CD30/TNFRSF8 (Phe19-Lys379) Accession # P29808.1 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Biotin |
| Species | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction