missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals IGF-1R Inhibitor, PPP

Product Code. 15437129
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437129 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15437129 Supplier Thermo Scientific Chemicals Supplier No. J65916.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 477-47-4 | C22H22O8, C22H22O8 | 414.41 g/mol

Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor
CAS 477-47-4
Molecular Formula C22H22O8
Formula Weight 414.40g/mol
Synonym Insulin-like Growth Factor-1 Receptor Inhibitor; Picropodophyllin
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.