Learn More
IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific Mouse anti-Rabbit, HRP, Secondary Antibody, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech SA00001-7L-1ML
Product Code. 17670307
  • 179.00 € / 1mL
Description
IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific Monoclonal antibody apecifically detetects IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific antigen in Rabbit samples. It is validated for ELSIA, Western Blot.
Specifications
Specifications
IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific
Monoclonal
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.
ELISA, Western Blot
HRP
Mouse
1 mL