missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Donkey anti-Mouse, CoraLite594, Secondary Antibody, Proteintech

Donkey anti-Mouse Affinipure Secondary Antibody

50.00 € - 99.00 €

Specifications

Antigen IgG(H+L)
Applications Immunofluorescence, Flow Cytometry
Conjugate CoraLite 594
Form Liquid
Host Species Donkey
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17630407
View Documents
Proteintech
SA00013-7-200UL
200 μL
50.00 €
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17640407
View Documents
Proteintech
SA00013-7-500UL
500 μL
99.00 €
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

IgG(H+L)
CoraLite 594
Donkey
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Immunofluorescence, Flow Cytometry
Liquid
Mouse
Secondary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.