Learn More
IgG(H+L) Donkey anti-Rabbit, CoraLite488, Secondary Antibody, Proteintech
Donkey anti-Rabbit Affinipure Secondary Antibody
Brand:  Proteintech SA00013-6-200UL
 View more versions of this product
Product Code. 17610407
  • 50.00 € / 200µL
Specifications
IgG(H+L)
CoraLite 488
Donkey
200 μL
Rabbit
Liquid
Immunofluorescence, Flow Cytometry
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.