Learn More
IgG(H+L) Donkey anti-Rat, AP, Secondary Antibody, Proteintech
Donkey anti-Rat Affinipure Secondary Antibody
Brand:  Proteintech SA00002-6-1ML
 View more versions of this product
Product Code. 17600327
  • 179.00 € / 1mL
Specifications
IgG(H+L)
Alkaline Phosphatase (AP)
Donkey
1 mL
Rat
Liquid
ELISA, Western Blot
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.