missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Goat anti-Mouse, CoraLite488, Secondary Antibody, Proteintech
Goat anti-Mouse Affinipure Secondary Antibody
50.00 € - 99.00 €
Specifications
| Antigen | IgG(H+L) |
|---|---|
| Applications | Immunofluorescence, Flow Cytometry |
| Conjugate | CoraLite 488 |
| Form | Liquid |
| Host Species | Goat |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17620397
|
Proteintech
SA00013-1-200UL |
200 μL |
50.00 €
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17630397
|
Proteintech
SA00013-1-500UL |
500 μL |
99.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| IgG(H+L) | |
| CoraLite 488 | |
| Goat | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
| Immunofluorescence, Flow Cytometry | |
| Liquid | |
| Mouse | |
| Secondary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title