Learn More
IgG(H+L) Goat anti-Swine, HRP, Secondary Antibody, Proteintech
Goat anti-Swine Affinipure Secondary Antibody
Brand:  Proteintech SA00001-5-1ML
Product Code. 17640307
  • 179.00 € / 1mL
Specifications
IgG(H+L)
HRP
Goat
1 mL
Swine
Liquid
ELISA, Western Blot
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.