Learn More
IgG(H+L) Rabbit anti-Goat, AMCA, Secondary Antibody, Proteintech
Rabbit anti-Goat Affinipure Secondary Antibody
Brand:  Proteintech SA00010-4-500UL
 View more versions of this product
Product Code. 17670377
  • 99.00 € / 500µL
Specifications
IgG(H+L)
AMCA
Rabbit
500 μL
Goat
Liquid
Immunofluorescence, Flow Cytometry
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.