Learn More
IgG(H+L) Rabbit anti-Goat, Cyanine3, Secondary Antibody, Proteintech
Rabbit anti-Goat Affinipure Secondary Antibody
Brand:  Proteintech SA00009-4-500UL
Product Code. 17690367
  • 99.00 € / 500µL
Specifications
IgG(H+L)
Cyanine3
Rabbit
500 μL
Goat
Liquid
Immunofluorescence, Flow Cytometry
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.