missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy PAX8 Ready-To-Use IHC Kit

Product Code. 17466743 Shop All Proteintech Products
Click to view available options
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17466743

Brand: Proteintech KHC003850T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A ready-to-use IHC kit for the staining of PAX8.

  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.

Specifications

Target Species Human, Rat, Mouse
Sample Type FFPE Tissue
Content And Storage Store at 2°C to 8°C
Packaging Quantity 50 Tests
Sufficient For 50 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.