missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Type III Soda Lime Glass Specimen Vial

Product Code. 14851582 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
2 mL
3 mL
4 mL
8 mL
16 mL
25 mL
35 mL
Unit Size:
Pack of 100
Pack of 250

Product Code. 14851582

Only null left
Add to basket
Edge
Add to basket

Use these dependable vials for small sample collection, analysis or storage. Fisherbrand™ Type III Soda Lime Glass Specimen Vials are ideal for a range of diagnostic, pharmaceutical and healthcare applications.

  • Clear Type III Soda Lime Glass vial
  • With push-in cap

Specifications

Diameter (Metric) Inner 25 mm
Color Clear
Material Soda Lime Glass (Type 3)
Volume (Metric) 16 mL
No. per Pack 100
Closure Material Polyethylene
Closure Type Push-in Cap
Quantity 100 Pk.
Height (Metric) 50 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.