Learn More
Stainless Steel Strap Clamp for IKA Eurostar Power Control-Visc P4 and P7 Stirrers
Product Code. 10618972 Shop All IKA Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
10618972 1 Each Each
1 options
Product Code. 10618972 Supplier IKA Supplier No. 0003159000
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use this stainless steel clamp for securing vessels against walls or synchronizing rotation during stirring. IKA™ Stainless Steel Strap Clamps are designed for IKA Eurostar Power Control-Visc P4 and P7 Stirrers
  • Includes cross collar joint for Boss Head 14-259-123 and fastening angle bar
  • For use with stands 14-259-86 and 14-259-039
  • Suitable for vessel from 1 3/8 to 11 in. (40 to 330mm) dia.
TRUSTED_SUSTAINABILITY
Specifications
Includes Boss Head Clamp R 270
Quantity 1 Each
Type RH 5 Strap Clamp