Learn More
IL-1 RI Mouse anti-Human, Clone: 732210, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB2691-SP
Encompass
Product Code. 15725524
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Human samples. It is validated for Western Blot.