missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20R alpha Mouse anti-Human, Clone: 173714, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB11762-SP
This item is not returnable.
View return policy
Description
IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| IL-20R alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9UHF4 | |
| IL20RA | |
| Mouse myeloma cell line NS0-derived recombinant human IL-20 R alpha Val30-Lys250 Accession # Q9UHF4 | |
| 25 μg | |
| Primary | |
| Detects human IL-20 R alpha in direct ELISAs and Western blots. In Western blots, approximately 5-15% cross-reactivity with recombinant human (rh) IFN-gamma R2, rhIL-10 R, rhIL-10 R beta , rhIL-20 R beta , and rhIL-22 BP is observed and no cross-reactivity with rhIFN-gamma RI, recombinant mouse IL-20 R alpha , or rhIL-22 R is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 173714 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20 R alpha, IL20R alpha, IL-20R1, IL-20R1IL-20 receptor subunit alpha, IL20RA, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 53832 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?