Learn More
IL-28R alpha/IFN-lambda R1 Sheep anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody has been used in 3 publications
Brand:  R&D Systems AF5260-SP
Encompass
Product Code. 15778562
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
IL-28R alpha/IFN-lambda R1 Polyclonal specifically detects IL-28R alpha/IFN-lambda R1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.