missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ExoProStar™ 1-step

Product Code. 11941411 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
100
500
2000
5000
Unit Size:
100 reactions
2000 reactions
500 reactions
5000 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
11941411 500 500 reactions
12124082 100 100 reactions
12134082 2000 2000 reactions
11961411 5000 5000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11941411 Supplier Cytiva Supplier No. US77705
  • ONLINE EXCLUSIVE PRICE
    471.55 €
    405.79 € / 500 reactions
    Save 65.76 €
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for enzymatic PCR clean-up. Cytiva Lifescience™ Illustra ExoProStar™ 1-step with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs.

  • One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
  • Just one pipetting step to set up your reaction
  • Fast 30-minute, hands-free protocol
  • 100% recovery of PCR product
  • Scalable for different reaction sizes
  • Automation friendly
TRUSTED_SUSTAINABILITY

Specifications

Description Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube.
Isolation Technology Enzymatic PCR Clean-up Technology
For Use With (Application) Remove unincorporated primers and dNTPs
No. of Reactions 500
Product Type illustra ExoProStar 1-Step
Quantity 1 Each
Test Time 30min.
Yield 100% recovery of PCR product
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.