missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ExoProStar™ 1-step

Use for enzymatic PCR clean-up.  Cytiva Lifescience™ Illustra ExoProStar™ 1-step  with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs.

Brand:  Cytiva US77702

99.10 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.



Code : GH

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 12124082

  • 108.93 € / 100 reactions
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
  • Just one pipetting step to set up your reaction
  • Fast 30-minute, hands-free protocol
  • 100% recovery of PCR product
  • Scalable for different reaction sizes
  • Automation friendly
Specifications

Specifications

Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube.
Remove unincorporated primers and dNTPs
illustra ExoProStar 1-Step
30min.
Enzymatic PCR Clean-up Technology
100
1 Each
100% recovery of PCR product
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers