Learn More
Cytiva Microspin G-50 Columns
Designed to allow the rapid purification of DNA for use in a wide range of applications using typical microfuge equipment. Cytiva illustra™ Microspin G-50 Columns will not remove or denature enzyme.
Brand: Cytiva 27-5330-02
860.19 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : GH
Additional Details : CAS Number : 9048-46-8 Weight : 0.88800kg
Description
- Optimal sample volumes in the range of 12-50μ
- For rapid purification of labeled DNA (>20 bases) from smaller molecules in solution
- Columns are prepacked with Sephadex G-50 Grade equilibrated in TE buffer with 0.05% Kathon CG/ICP Biocide
- illustra AutoSeq™ and illustra ProbeQuant™ G-50 Micro Columns are specifically designed for dye terminator removal and labelled probe purification respectively and offer optimized performance in those applications
- Can also be used for buffer exchange/desalting of enzymatic reactions (10 to 100 μL) following heat denaturation and phenol extraction
- Product recovery will be at least 10% lower than that from ProbeQuant G-50 Micro Columns.
- For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50 μl using spin-column chromatography
Desalting, buffer exchange; Removal of dye terminators from cycle sequencing reactions; Removal of labeled nucleotides from DNA labeling reactions
Specifications
Desalting, Buffer exchange, removal of dye terminators from cycle sequencing reactions, removal of labeled, Nucleotides in 25 to 50μL from DNA labeling reactions | |
Sephadex™ G-50 DNA grade | |
250 Preps | |
Column |