missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NAP™ Columns, NAP-5

Product Code. 10054394 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 Pack
50 Pack
Unit Size:
Pack of 20
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10054394 20 Pack Pack of 20
10218284 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10054394 Supplier Cytiva Supplier No. 17085301
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade

  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange

TRUSTED_SUSTAINABILITY

Specifications

Format Column
Isolation Technology Filtration
For Use With (Application) Rapid and efficient purification of DNA and Oligonucleotides
Includes Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
Product Type NAP-5
Quantity 20 Pack
compliance-icons
Product Identifier
  • illustra NAP 5 columns, (20)
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.