missing translation for 'onlineSavingsMsg'
Learn More

Cytiva plasmidPrep Mini Spin Kit

Product Code. 10379434 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
Sample Volume:
1.5 mL
3.0 mL
Unit Size:
50 reactions
Each
Product Code. No. of Reactions Sample Volume unitSize
10379434 250 Preps 1.5 mL Each
10772047 50 Preps 3.0 mL 50 reactions
2 options
This item is not returnable. View return policy

Product Code. 10379434

Brand: Cytiva 28904270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Spin columns prepacked with a novel silica-membrane; Suspension, lysis, and neutralization buffers containing chaotropic salts; Wash and elution buffers; Microcentrifuge collection tubes; Full protocol booklet; Detachable, quick reference protocol card

For the rapid isolation of high and low copy number plasmid DNA

The illustra plasmidPrep Mini Spin Kit uses a simple plasmid DNA purification protocol involving a modified alkaline lysis procedure and a novel silica-based membrane to achieve highly efficient plasmid DNA purification. Instead of organic solvents, the kit contains chaotropic salts to denature protein components and promote selective binding of DNA to the novel silica membrane.

  • Total preparation time of less than 10 minutes—representing a 50% reduction in protocol time
  • Produces high-quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing
  • Increased ease of use provided minimal changes in pipetting volume from one part of the protocol to the next, less centrifugation steps, color-coded kit components, and lack of organic solvents
  • The typical plasmid yields from a fresh 1.5 ml culture of E.coli containing a high copy number plasmid is approximately 9 μg
  • Can accommodate high and low copy number plasmid purification

TRUSTED_SUSTAINABILITY

Specifications

Description Purify high yields of plasmid DNA in less than 10 min. Color-coded tubes and columns simplify the protocol and purified DNA is suitable for all common downstream applications.
For Use With (Application) Suitable for many downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCRSequencing, PCR, ligation and cloning, restriction digest, enzymatic modification, template for labeled probe generation, transforma
Includes Spin columns prepacked with a novel Silica membrane, Suspension, Lysis and Neutralization Buffers containing chaotropic salts, Wash and Elution Buffers, microcentrifuge collection tubes, full protocol booklet, Detachable, quick reference protocol card
No. of Reactions 250 Preps
Product Type illustra plasmidPrep Mini Spin Kit
Purity 1.7
Sample Volume 1.5 mL
Test Time Approx. 9 min
Yield 6-15 μg
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium dodecyl sulphate, guanidinium chloride
  • UNKNOWN ACUTE-48.4 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 42% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.