missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PuReTaq Ready-To-Go™ PCR Beads

Product Code. 10525775 Shop All Cytiva Products
Change view
Click to view available options
Format:
0.2mL Tube with hinged cap
0.2mL Tubes/plate
0.5mL Tube
No. of Reactions:
96
100
480
Volume:
200μL
500μL
Unit Size:
100 reactions
480 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format No. of Reactions Volume Unit Size
10525775 0.2mL Tubes/plate 480 200μL 480 reactions
10380395 0.2mL Tubes/plate 96 200μL Each
10678095 0.2mL Tube with hinged cap 96 200μL Each
10289034 0.5mL Tube 100 500μL 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10525775 Supplier Cytiva Supplier No. 27955702
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-mixed, pre-dispensed reaction beads optimized for performing standard PCR amplifications. Cytiva Lifescience™ illustra™ PuReTaq Ready-To-Go™ PCR Beads are stable at ambient temperature.

  • puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
  • Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
  • Verified for real-time PCR
  • Ready-To-Go means simply add template DNA and primers and cycle

TRUSTED_SUSTAINABILITY

Specifica

Content And Storage Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP
Format 0.2mL Tubes/plate
Quantity 1 Kit
For Use With (Application) Standard PCR
No. of Reactions 480
Volume 200μL
Identificativo prodotto
  • PuReTaq
Indicazioni di pericolo
  • EUH210-Safety data sheet available on request.
Informazioni supplementari
  • UNKNOWN ACUTE-73.3 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 73.3% of components with unknown hazards to the aquatic environment.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.