missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Includes
- RAPD analysis beads, control E. coli BL21 DNA, control E. coli C1a DNA, and RAPD analysis primer 2. (DNA from two E. coli strains and RAPD analysis primer 2 are provided as controls.)
- The illustra RAPD Analysis Kit (Mfr. No. 27-9502-01) consists of the Ready-To-Go™ RAPD Analysis Beads and six lyophilized primers which can be used with RAPD Analysis Beads. Each primer in the set is an arbitrary 10-mer which is specifically designed for use in RAPD analysis.
- Preformulated, single-dose, ambient-temperature-stable reaction beads ensure greater reproducibility between reactions, minimize pipetting steps and reduce the potential for pipetting errors and contamination
- RAPD reactions are simple to perform—simply add genomic DNA solution and primer to a tube of RAPD Analysis Beads to a final reaction volume of 25 μL and cycle the reaction
- Each lot of RAPD Analysis Beads is function-tested to ensure its ability to generate a differential banding pattern between the two control E. coli strains using RAPD Analysis Primer 2 included with the RAPD beads
- RAPD reactions are preoptimized for use with a wide variety of organisms
- Beads are provided in thin-walled 0.5 mL tubes compatible with most thermal cyclers
Especificaciones
Especificaciones
| Content And Storage | Room temperature stable bead containing Buffer, dATP, dCTP, dGTP, dTTP, BSA, thermostable DNA Polymerases, control E. coli BL21(DE3) DNA: 1μg of E. coli BL21(DE3) DNA, lyophilized, control E. coli C1a DNA: 1μg of E. coli |
| Format | Tube |
| pH | 8.3 |
| Product Type | Analysis Beads |
| Quantity | 100 Reactions |
| For Use With (Application) | Standard PCR |
| Volume | 500 μL |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?