missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RNAspin Mini Isolation Kit

Extract high-quality RNA from a wide range of samples rapidly. Cytiva Life Sciences™illustra™ RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells, tissue, bacteria, or yeast.

Brand:  Cytiva 25-0500-70

Code : GE

Additional Details : CAS Number : 9048-46-8 Weight : 0.47300kg Transport : UN number : 1993 Chem class : 3 Pack group : III

 View more versions of this product

Product Code. 11535065

  • 147.02 € / 20 reactions
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Recovers total RNA due to removal of genomic DNA through on-column DNase I treatment
  • Can be used on cultured cells, tissue, yeast and hard-to-lyse bacterial strains
  • Prefilters reduce lysate viscosity for maximum yield and purity
  • Results can be obtained with small amounts of sample
  • Binding capacity: 100μg
  • Sample size: 30mg (animal tissue), up to 5 × 106 cells (cell culture)
  • Elution volume: 40 to 120μL
  • RNA Purity (A260/A280): 1.8 to 2.2
  • Total time of preparation: <30 min/6 preps

Quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments

Specifications

Specifications

illustra RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types.
Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I
illustra RNAspin Mini Kit
<30 mg
Up to 100μg
Used for the isolation of high-quality total RNA from a wide range of source material including hard-to-lyse bacterial strains. Purified RNA is suitable for use in sensitive downstream applications such as quantitative reverse transcriptase PCR (qRT-PCR),
20 Preps
1.8-2.2
< 30 min/6 preps
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers