missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios DETERG'ANIOS SURACTIF MARINE

Product Code. 11838871
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11838871 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11838871 Supplier Laboratoires Anios Supplier No. 785.209

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Daily detergent cleaning of all washable surfaces: floors (tiles, plastic coverings...), walls (tiles, washable paint...), toilets, metal-coated and plastic surfaces. Compatible when alternated with disinfectants.

Floors and surfaces Detergent:

  • Orange coloured solution
  • pH of straight product : approx. 10
  • pH of diluted product (0.25%) : approx. 7.6
  • Density at 20°C : approx. 1.028
  • Non corrosive (absence of oxidizing agents)

Specifications

For Use With (Application) Prepare circular DNA templates for cycle sequencing, cloning and transformation
Volume (Metric) 1 L
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: D-glucopyranose, oligomeriques, decyl octyl glycosides, isotridecanol ethoxylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.