missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Tissue and Cells genomicPrep Mini Spin Kit

Product Code. 10714547 Shop All Cytiva Products
Click to view available options
No. of Reactions:
250 Preps
50 Preps
Unit Size:
250 reactions
50 reactions
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10714547

missing translation for 'mfr': Cytiva 28904276

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Includes

Spin columns prepacked with a novel silica-membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; Full protocol booklet with a detachable, quick reference protocol card

Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures

To minimize the incidence of DNA shearing during extraction, the illustra tissue and cells genomicPrep Mini Spin Kit includes a lysis solution with proteinase K to gently release genomic DNA from tissues or cells. The kit can be used to extract DNA from diverse tissue types such as liver, kidney, and mouse tails with yields ranging from 0.5 to 1.5μg of genomic DNA per mg of tissue. The kit consistently generates purified genomic DNA with optical density A260/280 readings of about 1.8.

  • Capable of purifying up to 1.5μg/mg of tissue from an input tissue sample range of 5 to 50mg, and up to 40μg of genomic DNA per 5 million cells depending on cell type
  • Total DNA extraction time reduced significantly to 90 minutes (including tissue disruption)
  • Produces high-quality genomic DNA with high molecular weight and less degradation
  • Validated in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests

TRUSTED_SUSTAINABILITY

Especificaciones

Description illustra tissue and cells genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures in just 90 min for tissues and 45 min for ce
For Use With (Application) Used for several downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCR
Includes Spin columns prepacked with a novel Silica membrane, Lyophilized Proteinase K powder, Lysis solution, wash and Elution Buffers, microcentrifuge collection tubes, a full protocol booklet and a detachable, quick reference protocol card
No. of Reactions 250 Preps
Product Type illustra tissue and cells genomicPrep Mini Spin Kit
Purity 1.75
Sample Volume 5-50 mg
Test Time 90 min[Animal Tissue], 45 min[Cultured cells]
compliance-icons compliance-icons
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado