missing translation for 'onlineSavingsMsg'
Learn More

Iloprost, TRC

Product Code. 18083000
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083000 1 mg 1mg
18023300 10 mg 10mg
18080390 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083000 Supplier Toronto Research Chemicals Supplier No. TRCI2673001MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78919-13-8
Molecular Formula C22 H32 O4
Molecular Weight (g/mol) 360.49
Synonym Pentanoic acid, 5-[(3aS,4R,5R,6aS)-hexahydro-5-hydroxy-4-[(1E,3S)-3-hydroxy-4-methyl-1-octen-6-yn-1-yl]-2(1H)-pentalenylidene]-, (5E)-, Pentanoic acid, 5-[(3aS,4R,5R,6aS)-hexahydro-5-hydroxy-4-[(1E,3S)-3-hydroxy-4-methyl-1-octen-6-ynyl]-2(1H)-pentalenylidene]-, (5E)- (9CI), (5E)-5-[(3aS,4R,5R,6aS)-Hexahydro-5-hydroxy-4-[(1E,3S)-3-hydroxy-4-methyl-1-octen-6-yn-1-yl]-2(1H)-pentalenylidene]pentanoic acid, (1S,2R,3R,5S)-7-[(E)-4-Carboxybutylidene]-2-[(3S,1E)-3-hydroxy-4-methyl-6-octyn-1-enyl]-bicyclo[3.3.0]octane, Ciloprost, Endoprost, Ilomedin, Iloprost, ZK 36374
IUPAC Name (5E)-5-[(3aS,4R,5R,6aS)-5-hydroxy-4-[(E,3S)-3-hydroxy-4-methyloct-1-en-6-ynyl]-3,3a,4,5,6,6a-hexahydro-1H-pentalen-2-ylidene]pentanoic acid
SMILES CC#CCC(C)[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2C\C(=C/CCCC(=O)O)\C[C@H]12

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C22H32O4/c1-3-4-7-15(2)20(23)11-10-18-19-13-16(8-5-6-9-22(25)26)12-17(19)14-21(18)24/h8,10-11,15,17-21,23-24H,5-7,9,12-14H2,1-2H3,(H,25,26)/b11-10+,16-8+/t15?,17-,18+,19-,20+,21+/m0/s1
Formula Weight 360.2301
Percent Purity >95%
Chemical Name or Material Iloprost
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.