missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ LIVE Red and Green Caspase Apoptosis Detection Kits for microscopy

Product Code. 10700394
Change view
Click to view available options
Color:
Green, Red, Blue
Red, Green, Blue
Enzyme:
Caspase 3/7
Poly Caspases
Label or Dye:
FAM-DEVD-FMK, Propidium Iodide, Hoechst 33342
FAM-VAD-FMK, Propidium Iodide, Hoechst 33342
SR-DEVD-FMK, SYTOX Green, Hoechst 33342
SR-VAD-FMK, SYTOX Green, Hoechst 33342
Unit Size:
1 set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Enzyme Label or Dye unitSize
10700394 Red, Green, Blue Caspase 3/7 SR-DEVD-FMK, SYTOX Green, Hoechst 33342 1 set
10388932 Red, Green, Blue Poly Caspases SR-VAD-FMK, SYTOX Green, Hoechst 33342 1 set
10013272 Green, Red, Blue Poly Caspases FAM-VAD-FMK, Propidium Iodide, Hoechst 33342 1 set
10434402 Green, Red, Blue Caspase 3/7 FAM-DEVD-FMK, Propidium Iodide, Hoechst 33342 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10700394 Supplier Invitrogen™ Supplier No. I35102
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Detect apoptosis via microscopy, including fluorescence microscopy and HCS, with Image-iT LIVE Red and Green Caspase Apoptosis Detection Kits that report poly and caspase-3/7 activities.

Detect and evaluate caspase activation, membrane permeability, and cell cycle using Image-iT LIVE red and green poly and caspase-3 and-7 apoptosis detection kits for microscopy, including fluorescence microscopy and high content screening (HCS). The Image-iT LIVE red and green caspase detection kits use fluorescent inhibitor of caspases (FLICA) methodology to detect and report caspase activity, a measure of cell apoptosis. Specifically, the Image-iT LIVE red and green caspase detection kits contain the substrates SR-VAD-FMK and SR-DEVD-FMK (for red fluorescence), or FAM-VAD-FMK and FAM-DEVD-FMK (green fluorescence), for poly caspases and caspase-3/7 detection, respectively. They also include Hoechst 33342 and propidium iodide stains, enabling simultaneous evaluation of membrane permeability and cell cycle by microscopy and HCS.

The Image-iT LIVE red and green caspase detection kits for microscopy employ a novel approach to detecting active caspases: both assay kits take advantage of a fluorescent inhibitor of caspases (FLICA) methodology, which is an affinity label. This reagent associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. Different amino acid recognition sequences are used to detect different caspases: valine-alanine-aspartic acid (VAD) for poly-caspases (including caspase-1, -3, -4, -5, -6, -7, -8, and -9), and aspartic acid-glutamic acid-valine-aspartic acid (DEVD) for caspase 3/7. A sulforhodamine (SR) group or carboxyfluorescein (FAM) group is attached as a reporter.

The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining red or green fluorescent signal is a direct measure of the amount of active caspase present at the time the inhibitor was added. The approximate excitation and emission peaks of the FLICA, propidium iodide, and Hoechst 33342 reagents are 488⁄ nm/530 nm, 535⁄ nm/6617 nm, and 350 nm/⁄461 nm, respectively. The detection reagents included within the Image-iT LIVE Red Poly Caspases, Red Caspase-3/7, Green Poly Caspases, and Green Caspase-3/7 Detection kits are SR-VAD-FMK, SR-DEVD-FMK, FAM-VAD-FMK, and FAM-DEVD-FMK, respectively.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Description Image-iT LIVE Red Caspase-3 and -7 Detection Kit
Quantity 1 kit
Format Slide
Product Type Caspase 3/7 Reagent
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Excitation/Emission SR-DEVD-FMK: 550/595
SYTOX Green: 504/523
Hoechst 33342: 350/461
No. of Reactions 25 tests (labeling volumes of 300μL)
Color Red, Green, Blue
Shipping Condition Room Temperature
Product Line Image-iT
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Label or Dye SR-DEVD-FMK, SYTOX Green, Hoechst 33342
Label Type Other Label(s) or Dye(s)
Enzyme Caspase 3/7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.