missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Imatinib mesylate

Product Code. 18730298
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18730298 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18730298 Supplier R&D Systems™ Supplier No. 5906/100
  • ONLINE EXCLUSIVE PRICE
    188.26 €
    169.05 € / 100mg
    Save 19.21 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective v-Abl tyrosine kinase inhibitor; also inhibits PDGFR and c-kit

Imatinib mesylate is a potent and selective v-Abl tyrosine kinase inhibitor (IC50 = 38 nM). Also inhibits PDGFR and c-kit. Exhibits selectivity for v-Abl over a panel of other tyrosine and serine/threonine protein kinases. Selectively inhibits PDGF-stimulated growth of v-abl-transformed PB-3 cells and v-sis-transformed BALB/c 3T3 cells in vitro. Exhibits antitumor effects in mice bearing AMuLV or BABL/c 3T3 v-sis cells. Inhibits replication of SARS-CoV and MERS-CoV in vitro (EC50 values are 9.823 and 17.689 μM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide methanesulfonate
CAS 220127-57-1
Synonym Gleevec, CGP 57148B
Quantity 100 mg
Product Line Tocris
Target Abl Kinase Inhibitors
Molecular Formula C29H31N7O.CH4O3S
Formula Weight 589.71
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.