Learn More
Tocris Bioscience™ Imatinib mesylate
GREENER_CHOICE
Potent and selective v-Abl tyrosine kinase inhibitor; also inhibits PDGFR and c-kit
Brand:  Tocris Bioscience™ 5906/100
Encompass
Product Code. 15716795
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Imatinib mesylate is a potent and selective v-Abl tyrosine kinase inhibitor (IC50 = 38 nM). Also inhibits PDGFR and c-kit. Exhibits selectivity for v-Abl over a panel of other tyrosine and serine/threonine protein kinases. Selectively inhibits PDGF-stimulated growth of v-abl-transformed PB-3 cells and v-sis-transformed BALB/c 3T3 cells in vitro. Exhibits antitumor effects in mice bearing AMuLV or BABL/c 3T3 v-sis cells. Inhibits replication of SARS-CoV and MERS-CoV in vitro (EC50 values are 9.823 and 17.689 μM, respectively).
Specifications
Specifications
4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide methanesulfonate
220127-57-1
Gleevec, CGP 57148B