missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Imatinib (Mesylate)

Product Code. 17140082
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17140082 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17140082 Supplier STEMCELL Technologies Supplier No. 72532
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits ABL, PDGFR, and KIT

  • Imatinib mesylate is a first generation tyrosine kinase inhibitor that selectively targets certain tyrosine kinases, including ABL, platelet-derived growth factor receptor (PDGFR), and KIT (Druker, 2008; Müller)
  • Inhibits proliferation of primary cultured human mesenchymal stem cells, and promotes adipogenic over osteogenic differentiation (Fierro et al.)
  • Induces osteoblast differentiation in cultured osteoblastic cells, and reduces osteoclastogenesis in mouse bone marrow cultures (O'Sullivan et al.)
  • In CML, Imatinib inhibits the oncoprotein BCR-ABL, the product of the Philadelphia chromosome gene fusion (Carroll et al.; Druker et al.), 1996)
  • Inhibits autonomous erythropoiesis in peripheral blood mononuclear cells isolated from patients with polycythemia vera (Oehler et al.)

Specifications

CAS 220127-57-1
Synonym CGP57148B; Gleevec; Glivec; STI-571
Quantity 25 mg
Molecular Formula C29H31N7O · CH4SO3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.