missing translation for 'onlineSavingsMsg'
Learn More

Imatinib Mesylate, MedChemExpress

Product Code. 16481936
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481936 500 mg 500mg
16431336 200 mg 200mg
16430608 5 g 5g
16411959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481936 Supplier Medchem Express Supplier No. HY50946
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine kinases.

Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine kinases.

Specifications

Chemical Name or Material Imatinib Mesylate
CAS 220127-57-1
Color White
Purity Grade Notes Research
Molecular Formula C30H35N7O4S
Quantity 500 mg
Synonym STI571 MesylateCGP-57148B Mesylate
Solubility Information DMSO : 125 mg/mL (211.97 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (84.79 mM)
SMILES O=S(O)(C)=O.O=C(NC1=CC=C(C(NC2=NC=CC(C3=CC=CN=C3)=N2)=C1)C)C4=CC=C(CN5CCN(CC5)C)C=C4
Molecular Weight (g/mol) 589.71
Formula Weight 589.71
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.