missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ IMD 0354

Product Code. 18075492
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075492 10 mg 10mg
18066012 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075492 Supplier R&D Systems™ Supplier No. 2611/10
  • ONLINE EXCLUSIVE PRICE
    273.17 €
    245.70 € / 10mg
    Save 27.47 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of IKKβ

Inhibitor of IκB kinase-β (IKKβ) that blocks NF-κB nuclear translocation. Attenuates myocardial ischemia/reperfusion injury by decreasing expression of adhesion molecules ICAM-1 and P-selectin and inhibiting cytokine and chemokine production in cardiomyocytes. Induces G0/G1 cell cycle arrest and apoptosis in HMC-1 and breast cancer cells. Also antagonist at P2X1, P2X4 and P2X7 receptors (IC50 values are 19, 156 and 175 nM respectively). Inhibits platelet aggregation induced by collegen in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 978-62-1
Molecular Formula C15H8ClF6NO2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 383.67
Purity >99%
Molecular Weight (g/mol) 383.67
Chemical Name or Material IMD 0354
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.