missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Purmorphamine

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Purmorphamine is a smoothened (Smo) receptor agonist (EC50 ∼ 1 μM) and activates the Hedgehog pathway. Purmorphamine induces osteogenesis in mouse mesenchymal progenitor cells (C3H10T1/2). When combined with BMP-4, Purmorphamine can transdifferentiate pre-adipocytes (3T3-L1) and myoblasts (C2C12) into osteoblasts. Purmorphamine induces differentiation of multipotent mesenchymal progenitor cells into osteoblasts, and of spinal motor neurons from pluripotent human stem cells, and protects dopaminergic neurons in a mouse model of Parkinson's disease.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 520.62 |
| Molecular Weight (g/mol) | 520.637 |
| Chemical Name or Material | Purmorphamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction