missing translation for 'onlineSavingsMsg'
Learn More

Imidacloprid, MedChemExpress

Product Code. 16472216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472216 500 mg 500mg
16441446 200 mg 200mg
16418476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472216 Supplier Medchem Express Supplier No. HYB0838
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Imidacloprid is an effective and widely used neonicotinoid pesticide to control pests of cereals, vegetables, tea and cotton.

Imidacloprid is an effective and widely used neonicotinoid pesticide to control pests of cereals, vegetables, tea and cotton.

Specifications

Chemical Name or Material Imidacloprid
CAS 138261-41-3
Color White
Purity Grade Notes Research
Molecular Formula C9H10ClN5O2
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (391.14 mM) ∣H2O : 1 mg/mL (3.91 mM; ultrasonic and warming and heat to 80°C)
SMILES O=[N+](/N=C1NCCN\1CC2=CC=C(Cl)N=C2)[O-]
Molecular Weight (g/mol) 255.66
Formula Weight 255.66
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.