missing translation for 'onlineSavingsMsg'
Learn More

Imidacloprid, TRC

Product Code. 30394536
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394536 Supplier Toronto Research Chemicals Supplier No. TRCI2749905G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imidacloprid
CAS 138261-41-3
Purity Grade Notes HPLC
Molecular Formula C9 H10 Cl N5 O2
Quantity 5 g
Synonym 1-(6-Chlor-3-pyridinylmethyl)- N-nitroimidazolidin-2-ylidenamine, 2-Imidazolidinimine, 1-[(6-chloro-3-pyridinyl)methyl]-N-nitro-, (2E)-, (2E)-1-[(6-Chloro-3-pyridinyl)methyl]-N-nitro-2-imidazolidinimine, 1-(2-Chloro-5-pyridylmethyl)-2-(N-nitroimino)imidazolidine, 1-(6-Chloro-3-pyridylmethyl)-N-nitroimidazolidin-2-ylideneamine, 1-[(6-Chloro-3-pyridinyl)methyl]-4,5-dihydro-N-nitro-1H-imidazol-2-amine, 1-[(6-Chloro-3-pyridinyl)methyl]-N-nitro-2-imidazolidinimine, AE-F 106464-00GR01B0, AEF 106464, AGST 03001, Acceleron IX 409, Admire, Admire 2F, Admire Pro, Advantage Flea Adulticide, Advise, Alias, Alias 2F, BAY-NTN 33893, Baimieshi, Bayer Advanced Season-Long Grub control, Biotlin, Biunik 200SL, CP 1, Comodor, Confidate, Confidor, Confidor 100SL, Confidor 200 O-TEQ, Confidor 200SL, Confidor 240 O-TEQ, Confidor 350SC, Confidor 70WG, Confidor SL, CoreTect, Couraze, Couraze Max, Enforce, Enforce (pesticide), Gaucho, Gaucho 600FS, Gaucho Grande, Genesis, Genesis (pesticide), Grubex, Hachikusan, Imicide, Imidacloprid, Jade, Kohinor, Kumiai Admire, Lada, Lada (insecticide), Macho Max, Mallet 2F, Marathon, Marathon (insecticide), Marathon II, Merit, Merit (insecticide), Merit 2F, Meritgreen, NTN 33893, NTN 33893-240FS, NUP 06023, NUP 06024, NUQ 05054, Nuprid, Pasada, Premis, Premise, Premise (insecticide), Prescribe, Preventol TM, Pro-Agro, Provado, Provado 2F, Quick-Bayt, Rinfidor, Senator, Senator (neonicotinoid), Tanrec, Timbertreat N 98, Torrent, Trimax, Trimax (pesticide), Trimax Pro, Widow, Yashijing
InChI Formula InChI=1S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5H,3-4,6H2,(H,11,13)
SMILES [O-][N+](=O)\N=C\1/NCCN1Cc2ccc(Cl)nc2
IUPAC Name (NE)-N-[1-[(6-chloropyridin-3-yl)methyl]imidazolidin-2-ylidene]nitramide
Molecular Weight (g/mol) 255.66
Formula Weight 255.0523
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.