missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-2-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10127802
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10127802 250 mg 250mg
10578243 1 g 1g
10585285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127802 Supplier Thermo Scientific Maybridge Supplier No. CC31304CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 118000-43-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD06739247
InChI Key QKKKBOZQZHBENG-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde, 2-formylpyridoimidazole, 2,3-dihydroimidazo 1,2-a pyridine-2-carbaldehyde, acmc-20ac55, imidazo 1,2-apyridine-2-carbaldehyde, imid-azo 1,2-a pyridine-2-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde 9ci
PubChem CID 4961257
IUPAC Name imidazo[1,2-a]pyridine-2-carbaldehyde
SMILES C1=CC2=NC(=CN2C=C1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.