missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-3-carboxylic acid, 98%, BLD Pharm™

Product Code. 31110083
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31110083 250 mg
31108776 1 g
31103623 5 g
31096928 100 g
31100505 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31110083 Supplier BLD Pharm Supplier No. BD134402250mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6200-60-8
Molecular Formula C8H6N2O2
MDL Number MFCD01720458
Quantity 250 mg
SMILES O=C(C1=CN=C2C=CC=CN21)O
Molecular Weight (g/mol) 162.15
Percent Purity 98%
Chemical Name or Material Imidazo[1,2-a]pyridine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.